Assay Details
| Gene Symbol: DTX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981939.2 | XP_008980187.1 | 4 - 5 | 1335 | 91 | |
| XM_008981944.2 | XP_008980192.1 | 4 - 5 | 1335 | 91 | ||
| XM_008981955.2 | XP_008980203.1 | 5 - 6 | 1523 | 91 | ||
| XM_008981967.2 | XP_008980215.1 | 4 - 5 | 1180 | 91 | ||
| XM_008981969.2 | XP_008980217.1 | 6 - 7 | 1620 | 91 | ||
| XM_008981973.2 | XP_008980221.1 | 4 - 5 | 1400 | 91 | ||
| XM_008981974.2 | XP_008980222.1 | 5 - 6 | 1484 | 91 | ||
| XM_008981985.2 | XP_008980233.1 | 5 - 6 | 1422 | 91 | ||
| XM_008981989.2 | XP_008980237.1 | 3 - 4 | 1177 | 91 | ||
| XM_017966172.1 | XP_017821661.1 | 5 - 6 | 1239 | 91 | ||
More Information
Set Membership: |
|

DTX2
Genomic Map