Assay Details
| Gene Symbol: KLF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749702.4 | XP_002749748.1 | 1 - 2 | 757 | 78 | |
| XM_008999333.2 | XP_008997581.1 | 2 - 3 | 551 | 78 | ||
| XM_017973672.1 | XP_017829161.1 | 1 - 2 | 782 | 78 | ||
More Information
Set Membership: |
|

KLF7
Genomic Map