Assay Details
| Gene Symbol: KLHL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981302.2 | XP_008979550.1 | 2 - 3 | 3223 | 75 | |
| XM_008981303.2 | XP_008979551.1 | 3 - 4 | 1143 | 75 | ||
| XM_008981305.2 | XP_008979553.1 | 2 - 3 | 1080 | 75 | ||
| XM_008981308.2 | XP_008979556.1 | 2 - 3 | 3224 | 75 | ||
More Information
Set Membership: |
|

KLHL24
Genomic Map