Assay Details
| Gene Symbol: SLC7A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978145.1 | XP_017833634.1 | 3 - 4 | 2206 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC7A4| Gene Symbol: SLC7A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978145.1 | XP_017833634.1 | 3 - 4 | 2206 | 64 | |
Set Membership: |
|