Assay Details
| Gene Symbol: VIRMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759084.4 | XP_002759130.1 | 17 - 18 | 4304 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VIRMA| Gene Symbol: VIRMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759084.4 | XP_002759130.1 | 17 - 18 | 4304 | 107 | |
Set Membership: |
|