Assay Details
| Gene Symbol: BACE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967196.1 | XP_017822685.1 | 3 - 4 | 679 | 66 | |
| XM_017967197.1 | XP_017822686.1 | 3 - 4 | 679 | 66 | ||
More Information
Set Membership: |
|

BACE2
Genomic Map