Assay Details
| Gene Symbol: RDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009006217.2 | XP_009004465.1 | 6 - 7 | 804 | 109 | |
| XM_009006220.2 | XP_009004468.1 | 5 - 6 | 1765 | 109 | ||
| XM_017977493.1 | XP_017832982.1 | 5 - 6 | 1789 | 109 | ||
More Information
Set Membership: |
|

RDH12
Genomic Map