Assay Details
| Gene Symbol: LAMTOR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745555.4 | XP_002745601.1 | 3 - 4 | 293 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LAMTOR3| Gene Symbol: LAMTOR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745555.4 | XP_002745601.1 | 3 - 4 | 293 | 90 | |
Set Membership: |
|