Assay Details
| Gene Symbol: KANK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761694.3 | XP_002761740.2 | 7 - 8 | 2143 | 63 | |
| XM_008987137.2 | XP_008985385.1 | 7 - 8 | 2032 | 63 | ||
More Information
Set Membership: |
|

KANK3
Genomic Map