Assay Details
| Gene Symbol: CREB3L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751946.3 | XP_002751992.1 | 10 - 11 | 1688 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CREB3L2| Gene Symbol: CREB3L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751946.3 | XP_002751992.1 | 10 - 11 | 1688 | 62 | |
Set Membership: |
|