Assay Details
| Gene Symbol: NME4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755730.3 | XP_002755776.1 | 4 - 5 | 469 | 71 | |
| XM_009008827.2 | XP_009007075.1 | 3 - 4 | 367 | 71 | ||
| XM_009008828.1 | XP_009007076.1 | 4 - 5 | 672 | 71 | ||
More Information
Set Membership: |
|

NME4
Genomic Map