Assay Details
| Gene Symbol: MRPS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759546.4 | XP_002759592.2 | 7 - 8 | 997 | 80 | |
| XM_008983752.2 | XP_008982000.1 | 7 - 8 | 1049 | 80 | ||
| XM_008983754.2 | XP_008982002.1 | 7 - 8 | 1018 | 80 | ||
More Information
Set Membership: |
|

MRPS22
Genomic Map