Assay Details
| Gene Symbol: CRAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755759.4 | XP_002755805.3 | 3 - 4 | 698 | 74 | |
| XM_017978613.1 | XP_017834102.1 | 3 - 4 | 698 | 74 | ||
| XM_017978614.1 | XP_017834103.1 | 3 - 4 | 698 | 74 | ||
| XM_017978615.1 | XP_017834104.1 | 3 - 4 | 698 | 74 | ||
| XM_017978616.1 | XP_017834105.1 | 3 - 4 | 698 | 74 | ||
More Information
Set Membership: |
|

CRAMP1
Genomic Map