Assay Details
| Gene Symbol: RASL10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017964791.1 | XP_017820280.1 | 4 - 5 | 661 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RASL10A| Gene Symbol: RASL10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017964791.1 | XP_017820280.1 | 4 - 5 | 661 | 77 | |
Set Membership: |
|