Assay Details
| Gene Symbol: BCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978453.1 | XP_017833942.1 | 5 - 6 | 1392 | 59 | |
| XM_017978454.1 | XP_017833943.1 | 5 - 6 | 1458 | 59 | ||
More Information
Set Membership: |
|

BCR
Genomic Map