Assay Details
| Gene Symbol: LOC100393827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993187.2 | XP_008991435.1 | 3 - 4 | 1324 | 96 | |
| XM_017972017.1 | XP_017827506.1 | 3 - 4 | 242 | 96 | ||
More Information
Set Membership: |
|

LOC100393827
Genomic Map