Assay Details
| Gene Symbol: NECTIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762283.3 | XP_002762329.1 | 2 - 3 | 910 | 76 | |
| XM_017967653.1 | XP_017823142.1 | 2 - 3 | 910 | 76 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NECTIN2| Gene Symbol: NECTIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762283.3 | XP_002762329.1 | 2 - 3 | 910 | 76 | |
| XM_017967653.1 | XP_017823142.1 | 2 - 3 | 910 | 76 | ||