Assay Details
| Gene Symbol: CTDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963565.1 | XP_017819054.1 | 4 - 5 | 750 | 54 | |
| XM_017963566.1 | XP_017819055.1 | 4 - 5 | 750 | 54 | ||
| XM_017963567.1 | XP_017819056.1 | 4 - 5 | 403 | 54 | ||
| XM_017963568.1 | XP_017819057.1 | 4 - 5 | 2830 | 54 | ||
| XM_017963569.1 | XP_017819058.1 | 4 - 5 | 491 | 54 | ||
| XM_017963570.1 | XP_017819059.1 | 4 - 5 | 1822 | 54 | ||
| XM_017963571.1 | XP_017819060.1 | 2 - 3 | 454 | 54 | ||
| XM_017963572.1 | XP_017819061.1 | 4 - 5 | 461 | 54 | ||
More Information
Set Membership: |
|

CTDP1
Genomic Map