Assay Details
| Gene Symbol: OSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983424.2 | XP_008981672.1 | 3 - 4 | 3374 | 101 | |
| XM_017965667.1 | XP_017821156.1 | 3 - 4 | 970 | 101 | ||
More Information
Set Membership: |
|

OSR2
Genomic Map