Assay Details
| Gene Symbol: RAB11FIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756952.3 | XP_002756998.1 | 1 - 2 | 575 | 110 | |
| XM_008979321.2 | XP_008977569.1 | 1 - 2 | 575 | 110 | ||
More Information
Set Membership: |
|

RAB11FIP1
Genomic Map