Assay Details
| Gene Symbol: CD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267772.1 | NP_001254701.1 | 3 - 4 | 369 | 81 | |
| GenBank mRNA | AF452616.1 | - | 3 - 4 | 369 | 81 | |
More Information
Set Membership: |
|

CD4
Genomic Map