Assay Details
| Gene Symbol: LOC100408457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756759.4 | XP_002756805.1 | 1 - 2 | 68 | 78 | |
| GenBank mRNA | EU126881.1 | - | 1 - 2 | 74 | 78 | |
More Information
Set Membership: |
|

LOC100408457
Genomic Map