Assay Details
| Gene Symbol: MAPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757238.4 | XP_002757284.1 | 3 - 4 | 1702 | 72 | |
| XM_008979821.2 | XP_008978069.1 | 2 - 3 | 368 | 72 | ||
| XM_017963477.1 | XP_017818966.1 | 2 - 3 | 1809 | 72 | ||
More Information
Set Membership: |
|

MAPK4
Genomic Map