Assay Details
| Gene Symbol: ORMDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752587.4 | XP_002752633.1 | 4 - 5 | 447 | 76 | |
| XM_003733774.3 | XP_003733822.1 | 3 - 4 | 550 | 76 | ||
More Information
Set Membership: |
|

ORMDL2
Genomic Map