Assay Details
| Gene Symbol: LOC103790142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988847.2 | XP_008987095.1 | 3 - 4 | 366 | 89 | |
| XM_008988848.2 | XP_008987096.1 | 3 - 4 | 764 | 89 | ||
| XM_017967508.1 | XP_017822997.1 | 3 - 4 | 366 | 89 | ||
More Information
Set Membership: |
|

LOC103790142
Genomic Map