Assay Details
| Gene Symbol: FBXO31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761240.2 | XP_002761286.1 | 1 - 2 | 395 | 72 | |
| XM_008986333.2 | XP_008984581.1 | 1 - 2 | 395 | 72 | ||
| XM_017966813.1 | XP_017822302.1 | 1 - 2 | 395 | 72 | ||
More Information
Set Membership: |
|

FBXO31
Genomic Map