Assay Details
Gene Symbol: YPEL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002757882.3 | XP_002757928.2 | 2 - 3 | 731 | 68 | |
XM_008980910.1 | XP_008979158.1 | 2 - 3 | 477 | 68 | ||
XM_008980911.1 | XP_008979159.1 | 3 - 4 | 957 | 68 | ||
XM_008980914.1 | XP_008979162.1 | 3 - 4 | 722 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
