Assay Details
| Gene Symbol: RGS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747767.3 | XP_002747813.1 | 3 - 4 | 509 | 83 | |
| XM_008996300.2 | XP_008994548.1 | 3 - 4 | 336 | 83 | ||
| XM_008996301.2 | XP_008994549.1 | 2 - 3 | 411 | 83 | ||
| XM_017972177.1 | XP_017827666.1 | 2 - 3 | 2967 | 83 | ||
More Information
Set Membership: |
|

RGS19
Genomic Map