Assay Details
| Gene Symbol: IMPAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758924.4 | XP_002758970.1 | 1 - 2 | 663 | 79 | |
| XM_008982676.2 | XP_008980924.1 | 1 - 2 | 663 | 79 | ||
More Information
Set Membership: |
|

IMPAD1
Genomic Map