Assay Details
| Gene Symbol: C13H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757205.4 | XP_002757251.1 | 3 - 4 | 1207 | 63 | |
| XM_003734650.2 | XP_003734698.1 | 4 - 5 | 1227 | 63 | ||
| XM_008979740.2 | XP_008977988.1 | 4 - 5 | 1390 | 63 | ||
| XM_017963450.1 | XP_017818939.1 | 3 - 4 | 1044 | 63 | ||
More Information
Set Membership: |
|

C13H18orf25
Genomic Map