Assay Details
| Gene Symbol: EFNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984509.2 | XP_008982757.1 | 2 - 3 | 497 | 78 | |
| XM_017966324.1 | XP_017821813.1 | 3 - 4 | 1256 | 78 | ||
More Information
Set Membership: |
|

EFNA4
Genomic Map