Assay Details
| Gene Symbol: CYP2S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988059.2 | XP_008986307.2 | 7 - 8 | 1373 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP2S1| Gene Symbol: CYP2S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988059.2 | XP_008986307.2 | 7 - 8 | 1373 | 56 | |
Set Membership: |
|