Assay Details
| Gene Symbol: IGSF23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988241.2 | XP_008986489.2 | 5 - 6 | 2035 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IGSF23| Gene Symbol: IGSF23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988241.2 | XP_008986489.2 | 5 - 6 | 2035 | 90 | |
Set Membership: |
|