Assay Details
| Gene Symbol: COX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745740.4 | XP_002745786.2 | 2 - 3 | 578 | 86 | |
| XM_008993193.2 | XP_008991441.1 | 2 - 3 | 577 | 86 | ||
| XM_008993194.2 | XP_008991442.1 | 2 - 3 | 582 | 86 | ||
More Information
Set Membership: |
|

COX18
Genomic Map