Assay Details
| Gene Symbol: CD276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009005222.2 | XP_009003470.2 | 7 - 8 | 1661 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CD276| Gene Symbol: CD276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009005222.2 | XP_009003470.2 | 7 - 8 | 1661 | 65 | |
Set Membership: |
|