Assay Details
| Gene Symbol: CASP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745181.3 | XP_002745227.2 | 6 - 7 | 1857 | 65 | |
| XM_008992388.2 | XP_008990636.1 | 6 - 7 | 1739 | 65 | ||
| XM_008992389.2 | XP_008990637.1 | 6 - 7 | 1125 | 65 | ||
| XM_008992390.2 | XP_008990638.1 | 6 - 7 | 638 | 65 | ||
| XM_017970115.1 | XP_017825604.1 | 7 - 8 | 1848 | 65 | ||
More Information
Set Membership: |
|

CASP3
Genomic Map