Assay Details
| Gene Symbol: PUDP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762598.4 | XP_002762644.2 | 3 - 4 | 599 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PUDP| Gene Symbol: PUDP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762598.4 | XP_002762644.2 | 3 - 4 | 599 | 73 | |
Set Membership: |
|