Assay Details
| Gene Symbol: PLEKHG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992293.2 | XP_008990541.2 | 19 | 4937 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLEKHG4B| Gene Symbol: PLEKHG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992293.2 | XP_008990541.2 | 19 | 4937 | 98 | |
Set Membership: |
|