Assay Details
| Gene Symbol: LOC100406856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976345.1 | XP_017831834.1 | 1 | 124 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017976345.1 transcripts.
Set Membership: |
|

LOC100406856
Genomic Map