Assay Details
| Gene Symbol: WARS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754284.4 | XP_002754330.2 | 2 - 3 | 527 | 70 | |
| XM_009006551.2 | XP_009004799.1 | 3 - 4 | 373 | 70 | ||
| XM_009006552.2 | XP_009004800.1 | 3 - 4 | 584 | 70 | ||
| XM_009006553.2 | XP_009004801.1 | 4 - 5 | 473 | 70 | ||
| XM_009006554.2 | XP_009004802.1 | 2 - 3 | 710 | 70 | ||
| XM_017977333.1 | XP_017832822.1 | 3 - 4 | 345 | 70 | ||
More Information
Set Membership: |
|

WARS
Genomic Map