Assay Details
| Gene Symbol: WHAMM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998484.2 | XP_008996732.1 | 9 - 10 | 3246 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WHAMM| Gene Symbol: WHAMM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998484.2 | XP_008996732.1 | 9 - 10 | 3246 | 66 | |
Set Membership: |
|