Assay Details
| Gene Symbol: LRRC46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748543.3 | XP_002748589.1 | 6 - 7 | 865 | 53 | |
| GenBank mRNA | EF215102.1 | - | 2 - 3 | 117 | 53 | |
More Information
Set Membership: |
|

LRRC46
Genomic Map