Assay Details
| Gene Symbol: LOC100386005 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267770.1 | NP_001254699.1 | 566 | 81 | ||
| GenBank mRNA | EU041942.1 | - | 566 | 81 | ||
More Information
Set Membership: |
|

LOC100386005
Genomic Map