Assay Details
| Gene Symbol: PEA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760168.3 | XP_002760214.1 | 2 - 3 | 300 | 76 | |
| XM_008984713.1 | XP_008982961.1 | 2 - 3 | 366 | 76 | ||
| XM_017966399.1 | XP_017821888.1 | 2 - 3 | 194 | 76 | ||

PEA15
Genomic Map