Assay Details
| Gene Symbol: SPART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998133.2 | XP_008996381.2 | 4 - 5 | 1369 | 82 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPART| Gene Symbol: SPART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998133.2 | XP_008996381.2 | 4 - 5 | 1369 | 82 | |