Assay Details
| Gene Symbol: PTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751948.3 | XP_002751994.1 | 2 - 3 | 535 | 72 | |
| XM_009002999.1 | XP_009001247.1 | 2 - 3 | 535 | 72 | ||
More Information
Set Membership: |
|

PTN
Genomic Map