Assay Details
| Gene Symbol: IL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754412.3 | XP_002754458.1 | 4 - 5 | 447 | 100 | |
| XM_009006803.2 | XP_009005051.1 | 5 - 6 | 628 | 100 | ||
| XM_009006804.2 | XP_009005052.1 | 4 - 5 | 283 | 100 | ||
More Information
Set Membership: |
|

IL18
Genomic Map