Assay Details
Gene Symbol: LOC100407742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002743536.3 | XP_002743582.1 | 14 - 15 | 1860 | 65 | |
XM_009006649.2 | XP_009004897.1 | 14 - 15 | 1860 | 65 | ||
XM_009006658.2 | XP_009004906.1 | 13 - 14 | 1609 | 65 | ||
XM_009006666.2 | XP_009004914.1 | 13 - 14 | 3890 | 65 | ||
XM_017977894.1 | XP_017833383.1 | 8 - 9 | 985 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
