Assay Details
Gene Symbol: LOC100407742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002743536.3 | XP_002743582.1 | 17 - 18 | 2336 | 76 | |
XM_009006649.2 | XP_009004897.1 | 17 - 18 | 2336 | 76 | ||
XM_009006658.2 | XP_009004906.1 | 16 - 17 | 2085 | 76 | ||
XM_009006666.2 | XP_009004914.1 | 16 - 17 | 4366 | 76 | ||
XM_017977894.1 | XP_017833383.1 | 11 - 12 | 1461 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
