Assay Details
Gene Symbol: LOC100407742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002743536.3 | XP_002743582.1 | 4 - 5 | 550 | 109 | |
XM_009006649.2 | XP_009004897.1 | 4 - 5 | 550 | 109 | ||
XM_009006658.2 | XP_009004906.1 | 3 - 4 | 299 | 109 | ||
XM_009006666.2 | XP_009004914.1 | 3 - 4 | 2580 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009006658.2, XM_009006666.2 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
